Learn More
Analytical Sales & Services 58µL, 384-Well Collection Plate, Square Well / Spherical Bottom
Catalog No. 5000804467
Encompass
Advantage 58µL, 384-Well Collection Plate, Square Well / Spherical Bottom,  Polypropylene Plate\n\nQty/Pkg: 60
Catalog No. 50-008-04467 Supplier Analytical Sales & Services Supplier No. 38435
Advantage 58µL, 384-Well Collection Plate, Square Well / Spherical Bottom,  Polypropylene Plate\n\nQty/Pkg: 60