Learn More
Sino Biological Recombinant Mouse Ccl21a Protein
Supplier:  Sino Biological 58025MNAB100
Encompass_Preferred
A DNA sequence encoding the mouse CCL21A (NP_035254.1) (Met1-Gly133) was expressed.
Catalog No. 50-162-3195