Learn More
Sino Biological Recombinant Mouse GAS6 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 58026M08H100
Encompass_Preferred
A DNA sequence encoding the mouse GAS6 (NP_062394.2) (Met1-Pro674) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3197