Learn More
Sino Biological Recombinant Mouse Nectin-1/PVRL1 Protein (ECD, hFc Tag), HPLC-verified
Supplier:  Sino Biological 58137M02H100
Encompass_Preferred
A DNA sequence encoding the mouse NECTIN1 (NP_067399.2) (Met1-Ala354) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3199