Learn More
Eldex Laboratories Inc Preventive Maintenance Kit
Catalog No. NC0983414
Encompass_Preferred
Includes two graphite fiber reinforced PTFE piston seals, tools for installation, two valve cartridges, one retainer, a sintered stainless steel filter.
Catalog No. NC0983414 Supplier Eldex Laboratories Inc Supplier No. 5920
Includes two graphite fiber reinforced PTFE piston seals, tools for installation, two valve cartridges, one retainer, a sintered stainless steel filter.