Learn More
eMolecules​ 59247-47-1 | tert-Butyl 4-bromobenzoate | ChemScene | MFCD00093266 | 257.127 | C11H13BrO2 | 98.000 | CC(C)(C)OC(=O)c1ccc(Br)cc1 | 5g | 486155590
Catalog No. 5000490577 Shop All eMolecules Inc. Products
Encompass_Preferred
tert-Butyl 4-bromobenzoate | ChemScene | 59247-47-1 | MFCD00093266 | 257.127 | C11H13BrO2 | 98.000 | CC(C)(C)OC(=O)c1ccc(Br)cc1 | 5g | 486155590
Catalog No. 50-004-90577 Supplier eMolecules​ Supplier No. 486155590
tert-Butyl 4-bromobenzoate | ChemScene | 59247-47-1 | MFCD00093266 | 257.127 | C11H13BrO2 | 98.000 | CC(C)(C)OC(=O)c1ccc(Br)cc1 | 5g | 486155590