Learn More
ABclonal Technology 594 anti-Human CD303/BDCA-2
SDP
Catalog No. 5000009064
Encompass_Preferred
This gene encodes a member of the C-type lectin C-type lectin-like domain (CTL CTLD) superfamily Members of this family share a common protein fold and have diverse functions such as cell adhesion cell-cell signalling glycoprotein turnover and roles in inflammation and immune response The encoded type 2 transmembrane protein may play a role in dendritic cell function Two transcript variants encoding distinct isoforms have been identified for this gene
Catalog No. 50-000-09064 Supplier ABclonal Technology Supplier No. A24313200T
This gene encodes a member of the C-type lectin C-type lectin-like domain (CTL CTLD) superfamily Members of this family share a common protein fold and have diverse functions such as cell adhesion cell-cell signalling glycoprotein turnover and roles in inflammation and immune response The encoded type 2 transmembrane protein may play a role in dendritic cell function Two transcript variants encoding distinct isoforms have been identified for this gene