Learn More
ABclonal Technology 594 anti-Human CD66a/CEACAM1
SDP
Catalog No. 5000010605
Encompass_Preferred
This gene encodes a member of the carcinoembryonic antigen (CEA) family part of the immunoglobulin superfamily The CEA family includes two subgroups CEA cell adhesion molecules and pregnancy-specific glycoproteins located within a 1 2 Mb cluster on chromosome 19 The protein initially identified in liver bile ducts as biliary glycoprotein acts as a cell-cell adhesion molecule found on leukocytes epithelia and endothelia It mediates cell adhesion through both homophilic and heterophilic interactions with other proteins in the CEA subgroup The protein is involved in tissue differentiation angiogenesis apoptosis tumor suppression metastasis and immune response modulation Multiple transcript variants encode different isoforms but the full-length nature of all variants remains undefined
Catalog No. 50-000-10605 Supplier ABclonal Technology Supplier No. A24385200T
This gene encodes a member of the carcinoembryonic antigen (CEA) family part of the immunoglobulin superfamily The CEA family includes two subgroups CEA cell adhesion molecules and pregnancy-specific glycoproteins located within a 1 2 Mb cluster on chromosome 19 The protein initially identified in liver bile ducts as biliary glycoprotein acts as a cell-cell adhesion molecule found on leukocytes epithelia and endothelia It mediates cell adhesion through both homophilic and heterophilic interactions with other proteins in the CEA subgroup The protein is involved in tissue differentiation angiogenesis apoptosis tumor suppression metastasis and immune response modulation Multiple transcript variants encode different isoforms but the full-length nature of all variants remains undefined