Learn More
ABclonal Technology 594 p-PI3KP85a/B/P55Y
SDP
Catalog No. 5000014111
Encompass_Preferred
Phosphatidylinositol 3-kinase phosphorylates the inositol ring of phosphatidylinositol at the 3-prime position The enzyme comprises a 110 kD catalytic subunit and a regulatory subunit of either 85 55 or 50 kD This gene encodes the 85 kD regulatory subunit Phosphatidylinositol 3-kinase plays an important role in the metabolic actions of insulin and a mutation in this gene has been associated with insulin resistance Alternative splicing of this gene results in four transcript variants encoding different isoforms [provided by RefSeq Jun 2011]
Catalog No. 50-000-14111 Supplier ABclonal Technology Supplier No. AP1489100T
Phosphatidylinositol 3-kinase phosphorylates the inositol ring of phosphatidylinositol at the 3-prime position The enzyme comprises a 110 kD catalytic subunit and a regulatory subunit of either 85 55 or 50 kD This gene encodes the 85 kD regulatory subunit Phosphatidylinositol 3-kinase plays an important role in the metabolic actions of insulin and a mutation in this gene has been associated with insulin resistance Alternative splicing of this gene results in four transcript variants encoding different isoforms [provided by RefSeq Jun 2011]