Learn More
ABclonal Technology 594 Rabbit anti- CD226/DNAM-1
SDP
Catalog No. 5000009819
Encompass_Preferred
Predicted to enable integrin binding activity and protein kinase binding activity Acts upstream of or within positive regulation of interferon-gamma production and positive regulation of natural killer cell cytokine production Located in external side of plasma membrane Is expressed in embryo Orthologous to human CD226 (CD226 molecule)
Catalog No. 50-000-09819 Supplier ABclonal Technology Supplier No. A25864500T
Predicted to enable integrin binding activity and protein kinase binding activity Acts upstream of or within positive regulation of interferon-gamma production and positive regulation of natural killer cell cytokine production Located in external side of plasma membrane Is expressed in embryo Orthologous to human CD226 (CD226 molecule)