Learn More
ABclonal Technology 594 Rabbit anti FcRI/CD64
SDP
Catalog No. 5000005866
Encompass_Preferred
Enables IgG binding activity and IgG receptor activity Acts upstream of or within several processes including antibody-dependent cellular cytotoxicity endocytosis and positive regulation of hypersensitivity Located in external side of plasma membrane Is expressed in liver Orthologous to several human genes including FCGR1A (Fc gamma receptor Ia)
Catalog No. 50-000-05866 Supplier ABclonal Technology Supplier No. A23857200T
Enables IgG binding activity and IgG receptor activity Acts upstream of or within several processes including antibody-dependent cellular cytotoxicity endocytosis and positive regulation of hypersensitivity Located in external side of plasma membrane Is expressed in liver Orthologous to several human genes including FCGR1A (Fc gamma receptor Ia)