Learn More
ABclonal Technology 594 Rabbit anti-Human CD142
SDP
Catalog No. 5000007475
Encompass_Preferred
This gene encodes coagulation factor III a cell surface glycoprotein that initiates blood coagulation cascades by acting as a high-affinity receptor for factor VII The complex formed activates specific proteolysis initiating coagulation Unlike other cofactors factor III is fully functional when expressed on cell surfaces like monocytes It has extracellular transmembrane and cytoplasmic domains Platelets and monocytes express this factor under procoagulatory and proinflammatory conditions and it plays a role in HIV-associated coagulopathy Factor III expression in platelets and monocytes is linked to COVID-19 severity and mortality Congenital deficiency of this factor has not been observed Alternative splicing produces multiple transcript variants
Catalog No. 50-000-07475 Supplier ABclonal Technology Supplier No. A24183500T
This gene encodes coagulation factor III a cell surface glycoprotein that initiates blood coagulation cascades by acting as a high-affinity receptor for factor VII The complex formed activates specific proteolysis initiating coagulation Unlike other cofactors factor III is fully functional when expressed on cell surfaces like monocytes It has extracellular transmembrane and cytoplasmic domains Platelets and monocytes express this factor under procoagulatory and proinflammatory conditions and it plays a role in HIV-associated coagulopathy Factor III expression in platelets and monocytes is linked to COVID-19 severity and mortality Congenital deficiency of this factor has not been observed Alternative splicing produces multiple transcript variants