Learn More
ABclonal Technology 594 Rabbit anti-Human CD177
SDP
Supplier:  ABclonal Technology A24403500T
Encompass_Preferred
This gene encodes a glycosyl-phosphatidylinositol (GPI)-linked cell surface glycoprotein that plays a role in neutrophil activation The protein can bind platelet endothelial cell adhesion molecule-1 and function in neutrophil transmigration Mutations in this gene are associated with myeloproliferative diseases Over-expression of this gene has been found in patients with polycythemia rubra vera Autoantibodies against the protein may result in pulmonary transfusion reactions and it may be involved in Wegener s granulomatosis A related pseudogene which is adjacent to this gene on chromosome 19 has been identified
Catalog No. 50-000-09447