Learn More
ABclonal Technology 594 Rabbit anti-Human CD300a
SDP
Catalog No. 5000011303
Encompass_Preferred
This gene encodes a member of the CD300 glycoprotein family of cell surface proteins found on leukocytes involved in immune response signaling pathways This gene is located on chromosome 17 in a cluster with all but one of the other family members Multiple transcript variants encoding different isoforms have been found for this gene
Catalog No. 50-000-11303 Supplier ABclonal Technology Supplier No. A25743200T
This gene encodes a member of the CD300 glycoprotein family of cell surface proteins found on leukocytes involved in immune response signaling pathways This gene is located on chromosome 17 in a cluster with all but one of the other family members Multiple transcript variants encoding different isoforms have been found for this gene