Learn More
ABclonal Technology 594 Rabbit anti-Human CD307b
SDP
Catalog No. 5000011710
Encompass_Preferred
This gene encodes a member of the immunoglobulin receptor superfamily and is one of several Fc receptor-like glycoproteins clustered on the long arm of chromosome 1 The encoded protein has four extracellular C2-type immunoglobulin domains a transmembrane domain and a cytoplasmic domain that contains one immunoreceptor-tyrosine activation motif and two immunoreceptor-tyrosine inhibitory motifs This protein may be a prognostic marker for chronic lymphocytic leukemia Alternatively spliced transcript variants have been described but their biological validity has not been determined
Catalog No. 50-000-11710 Supplier ABclonal Technology Supplier No. A25525200T
This gene encodes a member of the immunoglobulin receptor superfamily and is one of several Fc receptor-like glycoproteins clustered on the long arm of chromosome 1 The encoded protein has four extracellular C2-type immunoglobulin domains a transmembrane domain and a cytoplasmic domain that contains one immunoreceptor-tyrosine activation motif and two immunoreceptor-tyrosine inhibitory motifs This protein may be a prognostic marker for chronic lymphocytic leukemia Alternatively spliced transcript variants have been described but their biological validity has not been determined