Learn More
ABclonal Technology 594 Rabbit anti-Human CD47
Supplier: ABclonal Technology A24302500T
This gene encodes a membrane protein which is involved in the increase in intracellular calcium concentration that occurs upon cell adhesion to extracellular matrix The encoded protein is also a receptor for the C-terminal cell binding domain of thrombospondin and it may play a role in membrane transport and signal transduction This gene has broad tissue distribution and is reduced in expression on Rh erythrocytes Alternatively spliced transcript variants have been found for this gene