Learn More
ABclonal Technology 594 Rabbit anti-Human CD55
SDP
Catalog No. 5000011182
Encompass_Preferred
This gene encodes a glycoprotein involved in the regulation of the complement cascade Binding of the encoded protein to complement proteins accelerates their decay thereby disrupting the cascade and preventing damage to host cells Antigens present on this protein constitute the Cromer blood group system (CROM) Alternative splicing results in multiple transcript variants The predominant transcript variant encodes a membrane-bound protein but alternatively spliced transcripts may produce soluble proteins
Catalog No. 50-000-11182 Supplier ABclonal Technology Supplier No. A24792100T
This gene encodes a glycoprotein involved in the regulation of the complement cascade Binding of the encoded protein to complement proteins accelerates their decay thereby disrupting the cascade and preventing damage to host cells Antigens present on this protein constitute the Cromer blood group system (CROM) Alternative splicing results in multiple transcript variants The predominant transcript variant encodes a membrane-bound protein but alternatively spliced transcripts may produce soluble proteins