Learn More
ABclonal Technology 594 Rabbit anti-Human CEACAM6
SDP
Catalog No. 5000006657
Encompass_Preferred
This gene encodes a protein that belongs to the carcinoembryonic antigen (CEA) family whose members are glycosyl phosphatidyl inositol (GPI) anchored cell surface glycoproteins Members of this family play a role in cell adhesion and are widely used as tumor markers in serum immunoassay determinations of carcinoma This gene affects the sensitivity of tumor cells to adenovirus infection The protein encoded by this gene acts as a receptor for adherent-invasive E coli adhesion to the surface of ileal epithelial cells in patients with Crohn s disease This gene is clustered with genes and pseudogenes of the cell adhesion molecules subgroup of the CEA family on chromosome 19
Catalog No. 50-000-06657 Supplier ABclonal Technology Supplier No. A24188100T
This gene encodes a protein that belongs to the carcinoembryonic antigen (CEA) family whose members are glycosyl phosphatidyl inositol (GPI) anchored cell surface glycoproteins Members of this family play a role in cell adhesion and are widely used as tumor markers in serum immunoassay determinations of carcinoma This gene affects the sensitivity of tumor cells to adenovirus infection The protein encoded by this gene acts as a receptor for adherent-invasive E coli adhesion to the surface of ileal epithelial cells in patients with Crohn s disease This gene is clustered with genes and pseudogenes of the cell adhesion molecules subgroup of the CEA family on chromosome 19