Learn More
ABclonal Technology 594 Rabbit anti-Human DLL4
Supplier: ABclonal Technology A25867100T
This gene is a homolog of the Drosophila delta gene The delta gene family encodes Notch ligands that are characterized by a DSL domain EGF repeats and a transmembrane domain