Learn More
ABclonal Technology 594 Rabbit anti-Human FAP
SDP
Catalog No. 5000011274
Encompass_Preferred
The protein encoded by this gene is a homodimeric integral membrane gelatinase belonging to the serine protease family It is selectively expressed in reactive stromal fibroblasts of epithelial cancers granulation tissue of healing wounds and malignant cells of bone and soft tissue sarcomas This protein is thought to be involved in the control of fibroblast growth or epithelial-mesenchymal interactions during development tissue repair and epithelial carcinogenesis Alternatively spliced transcript variants encoding different isoforms have been found for this gene
Catalog No. 50-000-11274 Supplier ABclonal Technology Supplier No. A24967100T
The protein encoded by this gene is a homodimeric integral membrane gelatinase belonging to the serine protease family It is selectively expressed in reactive stromal fibroblasts of epithelial cancers granulation tissue of healing wounds and malignant cells of bone and soft tissue sarcomas This protein is thought to be involved in the control of fibroblast growth or epithelial-mesenchymal interactions during development tissue repair and epithelial carcinogenesis Alternatively spliced transcript variants encoding different isoforms have been found for this gene