Learn More
ABclonal Technology 594 Rabbit anti-Human FATP1
Supplier: ABclonal Technology A25572100T
Enables biotin transmembrane transporter activity efflux transmembrane transporter activity and long-chain fatty acid transporter activity Involved in several processes including carboxylic acid transmembrane transport glycerophospholipid biosynthetic process and lipid transport across blood-brain barrier Located in membrane Part of plasma membrane