Learn More
ABclonal Technology 594 Rabbit anti-Human FceR1a
Supplier: ABclonal Technology A25412100T
The immunoglobulin epsilon receptor (IgE receptor) is the initiator of the allergic response When two or more high-affinity IgE receptors are brought together by allergen-bound IgE molecules mediators such as histamine that are responsible for allergy symptoms are released This receptor is comprised of an alpha subunit a beta subunit and two gamma subunits The protein encoded by this gene represents the alpha subunit