Learn More
ABclonal Technology 594 Rabbit anti-Human IL-5
Supplier: ABclonal Technology A24235500T
This gene encodes a cytokine that acts as a growth and differentiation factor for both B cells and eosinophils The encoded cytokine plays a major role in the regulation of eosinophil formation maturation recruitment and survival The increased production of this cytokine may be related to pathogenesis of eosinophil-dependent inflammatory diseases This cytokine functions by binding to its receptor which is a heterodimer whose beta subunit is shared with the receptors for interleukine 3 (IL3) and colony stimulating factor 2 (CSF2 GM-CSF) This gene is located on chromosome 5 within a cytokine gene cluster which includes interleukin 4 (IL4) interleukin 13 (IL13) and CSF2 This gene IL4 and IL13 may be regulated coordinately by long-range regulatory elements spread over 120 kilobases on chromosome 5q31