Learn More
ABclonal Technology 594 Rabbit anti-Human IL23R
Supplier: ABclonal Technology A25556200T
The protein encoded by this gene is a subunit of the receptor for IL23A IL23 This protein pairs with the receptor molecule IL12RB1 IL12Rbeta1 and both are required for IL23A signaling This protein associates constitutively with Janus kinase 2 (JAK2) and also binds to transcription activator STAT3 in a ligand-dependent manner