Learn More
ABclonal Technology 594 Rabbit anti-Human IL23R
SDP
Catalog No. 5000011560
Encompass_Preferred
The protein encoded by this gene is a subunit of the receptor for IL23A IL23 This protein pairs with the receptor molecule IL12RB1 IL12Rbeta1 and both are required for IL23A signaling This protein associates constitutively with Janus kinase 2 (JAK2) and also binds to transcription activator STAT3 in a ligand-dependent manner
Catalog No. 50-000-11560 Supplier ABclonal Technology Supplier No. A25556500T
The protein encoded by this gene is a subunit of the receptor for IL23A IL23 This protein pairs with the receptor molecule IL12RB1 IL12Rbeta1 and both are required for IL23A signaling This protein associates constitutively with Janus kinase 2 (JAK2) and also binds to transcription activator STAT3 in a ligand-dependent manner