Learn More
ABclonal Technology 594 Rabbit anti-Human IL9
Supplier: ABclonal Technology A24307200T
The protein encoded by this gene is a cytokine that acts as a regulator of a variety of hematopoietic cells This cytokine stimulates cell proliferation and prevents apoptosis It functions through the interleukin 9 receptor (IL9R) which activates different signal transducer and activator (STAT) proteins and thus connects this cytokine to various biological processes The gene encoding this cytokine has been identified as a candidate gene for asthma Genetic studies on a mouse model of asthma demonstrated that this cytokine is a determining factor in the pathogenesis of bronchial hyperresponsiveness