Learn More
ABclonal Technology 594 Rabbit anti-Human LAMP2
Supplier: ABclonal Technology A24189100T
The protein encoded by this gene is a member of a family of membrane glycoproteins This glycoprotein provides selectins with carbohydrate ligands It may play a role in tumor cell metastasis It may also function in the protection maintenance and adhesion of the lysosome Alternative splicing of this gene results in multiple transcript variants encoding distinct proteins