Learn More
ABclonal Technology 594 Rabbit anti-Human ROR1
SDP
Supplier:  ABclonal Technology A24902500T
Encompass_Preferred
This gene encodes a receptor tyrosine kinase-like orphan receptor that modulates neurite growth in the central nervous system The encoded protein is a glycosylated type I membrane protein that belongs to the ROR subfamily of cell surface receptors It is a pseudokinase that lacks catalytic activity and may interact with the non-canonical Wnt signalling pathway This gene is highly expressed during early embryonic development but expressed at very low levels in adult tissues Increased expression of this gene is associated with B-cell chronic lymphocytic leukaemia Alternative splicing results in multiple transcript variants encoding different isoforms
Catalog No. 50-000-08616