Learn More
Cayman Chemical 5α,6β-Dihydroxycholestanol, Purity: ≥95%, CAS: 1253-84-5, 10 mg
SDP
Catalog No. 503015939
Encompass_Preferred
An oxysterol metabolite of cholesterol; inhibits NMDA-mediated calcium influx in HEK293 cells expressing NR1/NR2B NMDA receptors; binds to Nav channels and decreases action potentials in hippocampal neurons in vitro at 10 µM; increases survival of spinal cord motoneurons, cortical neurons, and cerebellar granule neurons in vitro at 5-15 µM; neuroprotective in a rat model of cerebral ischemia at 12 mg/kg ; increases latency to seizure onset and reduces severity of PTZ-induced seizures in rats; has been used as a replacement for cholesterol in the study of cholesterol binding proteins
Catalog No. 50-301-5939 Supplier Cayman Chemical Supplier No. 2553810 MG
An oxysterol metabolite of cholesterol; inhibits NMDA-mediated calcium influx in HEK293 cells expressing NR1/NR2B NMDA receptors; binds to Nav channels and decreases action potentials in hippocampal neurons in vitro at 10 µM; increases survival of spinal cord motoneurons, cortical neurons, and cerebellar granule neurons in vitro at 5-15 µM; neuroprotective in a rat model of cerebral ischemia at 12 mg/kg ; increases latency to seizure onset and reduces severity of PTZ-induced seizures in rats; has been used as a replacement for cholesterol in the study of cholesterol binding proteins