Learn More
Cayman Chemical 5a 6a-epoxy Cholestnol 100mg
SDP
Catalog No. 503015917
Encompass_Preferred
An oxysterol and an oxidative metabolite of cholesterol; accumulates in MCF-7 breast cancer cells following tamoxifen and PBPE application and induces triacylglycerol biosynthesis by binding to LXRβ; levels are increased in rat aorta and mesenteric artery following orchidectomy; levels are increased in post-mortem frontal and occipital cortex of patients with Alzheimer’s disease
Catalog No. 50-301-5917 Supplier Cayman Chemical Supplier No. 25539100 MG
An oxysterol and an oxidative metabolite of cholesterol; accumulates in MCF-7 breast cancer cells following tamoxifen and PBPE application and induces triacylglycerol biosynthesis by binding to LXRβ; levels are increased in rat aorta and mesenteric artery following orchidectomy; levels are increased in post-mortem frontal and occipital cortex of patients with Alzheimer’s disease