Learn More
Cayman Chemical 5α,6α-epoxy Cholestanol, Purity: ≥95%, CAS: 1250-95-9, 250 mg
SDP
Catalog No. 503015953
Encompass_Preferred
An oxysterol and an oxidative metabolite of cholesterol; accumulates in MCF-7 breast cancer cells following tamoxifen and PBPE application and induces triacylglycerol biosynthesis by binding to LXRβ; levels are increased in rat aorta and mesenteric artery following orchidectomy; levels are increased in post-mortem frontal and occipital cortex of patients with Alzheimer’s disease
Catalog No. 50-301-5953 Supplier Cayman Chemical Supplier No. 25539250 MG
An oxysterol and an oxidative metabolite of cholesterol; accumulates in MCF-7 breast cancer cells following tamoxifen and PBPE application and induces triacylglycerol biosynthesis by binding to LXRβ; levels are increased in rat aorta and mesenteric artery following orchidectomy; levels are increased in post-mortem frontal and occipital cortex of patients with Alzheimer’s disease