Learn More
Apexbio Technology LLC 5A-CHOLESTAN-3-ONE-250mg
SDP
Catalog No. 5000897547
Encompass_Preferred
5 -Cholestan-3-one (CAS No 566-88-1) is a steroidal compound often studied in biomedical research for its involvement in cholesterol metabolism pathways Originating from the cholestane family it may interact with enzymes that regulate lipid biosynthesis Its activity is typically analyzed in vitro with efficacies commonly observed in the nanomolar to low micromolar range 5 -Cholestan-3-one is utilized in various applications including studies using cell models and animal experiments and plays a role in drug screening processes Experimental concentrations typically depend on the specific research design
Catalog No. 50-008-97547 Supplier Apexbio Technology LLC Supplier No. C6621250
5 -Cholestan-3-one (CAS No 566-88-1) is a steroidal compound often studied in biomedical research for its involvement in cholesterol metabolism pathways Originating from the cholestane family it may interact with enzymes that regulate lipid biosynthesis Its activity is typically analyzed in vitro with efficacies commonly observed in the nanomolar to low micromolar range 5 -Cholestan-3-one is utilized in various applications including studies using cell models and animal experiments and plays a role in drug screening processes Experimental concentrations typically depend on the specific research design