Learn More
Cayman Chemical 5α-Cholestane, Purity: ≥95%, CAS: 481-21-0, 250 mg
SDP
Catalog No. 502934954
Encompass_Preferred
A sterol; has been found in dust samples from urban and rural paved and agricultural and public unpaved roads; has been used as an internal standard for the quantification of phytosterols by HPLC-MS/MS and fecal sterols by GC-FID and GC-MS
Catalog No. 50-293-4954 Supplier Cayman Chemical Supplier No. 26763250 MG
A sterol; has been found in dust samples from urban and rural paved and agricultural and public unpaved roads; has been used as an internal standard for the quantification of phytosterols by HPLC-MS/MS and fecal sterols by GC-FID and GC-MS