Learn More
Cayman Chemical 5α-hydroxy Laxogenin, Purity: ≥95%, CAS: 56786-63-1, 100 mg
SDP
Catalog No. 502937087
Encompass_Preferred
A brassinosteroid analog and a derivative of diosgenin; topical administration in combination with a commercial fertilizer increases the yield and fresh weight of endives (C. endivia) at 4, 8, and 12 ppm; inhibits sodium chloride-induced decreases in the fresh weight of lettuce shoots and roots when applied topically at 0.1 and 1 μM, as well as inhibits increases in ethylene emission at 1 μM
Catalog No. 50-293-7087 Supplier Cayman Chemical Supplier No. 27665100 MG
A brassinosteroid analog and a derivative of diosgenin; topical administration in combination with a commercial fertilizer increases the yield and fresh weight of endives (C. endivia) at 4, 8, and 12 ppm; inhibits sodium chloride-induced decreases in the fresh weight of lettuce shoots and roots when applied topically at 0.1 and 1 μM, as well as inhibits increases in ethylene emission at 1 μM