Learn More
Cayman Chemical 5a-ndrost-16-en-3-on 10mg
SDP
Catalog No. 502968139
Encompass_Preferred
A mammalian pheromone used to prime sexual behavior of sows in estrus for mating or artificial insemination; also used to study receptor-mediated odorant detection and the genetic basis for anosmias
Catalog No. 50-296-8139 Supplier Cayman Chemical Supplier No. 1823310 MG
A mammalian pheromone used to prime sexual behavior of sows in estrus for mating or artificial insemination; also used to study receptor-mediated odorant detection and the genetic basis for anosmias