Learn More
Cayman Chemical 5α-Androst-16-en-3α-ol, Purity: ≥95%, CAS: 1153-51-1, 50 mg
SDP
Catalog No. 502950119
Encompass_Preferred
A pheromone with diverse biological activities; enhances GABA-activated currents in primary mouse cerebellar granule cells (EC50 = 0.4 µM); increases the amplitude of GABA-activated currents in HEK293 cells expressing human α1β2γ2 and α2β2γ2 subunit-containing GABAA receptors from 0.1-1 µM; has antidepressant-like effects in mice from 5-10 mg/kg; has anxiolytic-like effects in mice from 30-50 mg/kg; protects against PTZ- or electroshock-induced seizures in mice (ED50s = 48.9 and 21.9 mg/kg, respectively)
Catalog No. 50-295-0119 Supplier Cayman Chemical Supplier No. 3345750 MG
A pheromone with diverse biological activities; enhances GABA-activated currents in primary mouse cerebellar granule cells (EC50 = 0.4 µM); increases the amplitude of GABA-activated currents in HEK293 cells expressing human α1β2γ2 and α2β2γ2 subunit-containing GABAA receptors from 0.1-1 µM; has antidepressant-like effects in mice from 5-10 mg/kg; has anxiolytic-like effects in mice from 30-50 mg/kg; protects against PTZ- or electroshock-induced seizures in mice (ED50s = 48.9 and 21.9 mg/kg, respectively)