Learn More
Cayman Chemical 5a-Pregn-3a 20a-dIol 10mg
SDP
Catalog No. 502918882
Encompass_Preferred
An active metabolite of progesterone; increases intracellular calcium levels in washed isolated human platelets at 10 µM
Catalog No. 50-291-8882 Supplier Cayman Chemical Supplier No. 3889010 MG
An active metabolite of progesterone; increases intracellular calcium levels in washed isolated human platelets at 10 µM