Learn More
Sino Biological Recombinant Mouse PLA2G2D Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 5A1107M02H100
Encompass_Preferred
A DNA sequence encoding the mouse PLA2G2D (NP_035239.1) (Met1-Cys144) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3203