Learn More
Medchemexpress LLC 5A2-SC8 TFA | 1857341-90-2 | 98.6% | C93H173N5O20S5.xC2HF3O2 | 1 MG
SDP
Catalog No. 5000354303
Encompass_Preferred
5A2-SC8 TFA is an ionizable amino lipid utilized in lipid nanoparticles (LNPs), demonstrating significant potential for drug delivery with low in vivo toxicity. It facilitates the efficient delivery of small RNAs, such as siRNA and miRNA, specifically into tumor cells. This compound's LNPs can uniquely influence the delivery fate of RNA within the liver, thereby improving therapeutic outcomes in various cancer models.
  • High delivery potential.
  • Low in vivo toxicity.
  • Enables efficient delivery of small RNAs (siRNA and miRNA) into tumor cells.
  • Can alter therapeutic outcomes in cancer models by influencing RNA delivery in the liver.
Catalog No. 50-003-54303 Supplier Medchemexpress LLC Supplier No. HY145799A1MG
5A2-SC8 TFA is an ionizable amino lipid utilized in lipid nanoparticles (LNPs), demonstrating significant potential for drug delivery with low in vivo toxicity. It facilitates the efficient delivery of small RNAs, such as siRNA and miRNA, specifically into tumor cells. This compound's LNPs can uniquely influence the delivery fate of RNA within the liver, thereby improving therapeutic outcomes in various cancer models.
  • High delivery potential.
  • Low in vivo toxicity.
  • Enables efficient delivery of small RNAs (siRNA and miRNA) into tumor cells.
  • Can alter therapeutic outcomes in cancer models by influencing RNA delivery in the liver.