Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5alpha-Cholestan-3-one, 97%

Catalog No. AAL0872606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0872606 5 g
AAL0872603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0872606 Supplier Thermo Scientific Chemicals Supplier No. L0872606
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 566-88-1 | C27H46O | 386.664 g/mol

It is employed as a intermediate for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for organic synthesis.

Solubility
Insoluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 566-88-1
Molecular Formula C27H46O
Molecular Weight (g/mol) 386.664
MDL Number MFCD00065901
InChI Key PESKGJQREUXSRR-UXIWKSIVSA-N
Synonym 5alpha-cholestan-3-one, 5a-cholestan-3-one, 5alpha-cholestanone, coprostanone, 5alpha-cholestane-3-one, cholestan-3-one, 5a, 3-keto-5alpha-cholestane, 5-alpha-cholestan-3-one, cholestan-3-one
PubChem CID 92128
ChEBI CHEBI:17762
IUPAC Name (5S,8R,9S,10S,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-1,2,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydrocyclopenta[a]phenanthren-3-one
SMILES CC(C)CCCC(C)C1CCC2C1(CCC3C2CCC4C3(CCC(=O)C4)C)C

Specifications

Melting Point 128°C to 132°C
Quantity 5 g
Beilstein 2625580
Solubility Information Insoluble in water.
Optical Rotation +43° (c=1 in Chloroform)
Formula Weight 386.66
Percent Purity 97%
Chemical Name or Material 5alpha-Cholestan-3-one
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.