Learn More
Cayman Chemical 5-bromo-3-indolyl-β-D-Galactopyranoside, Purity: ≥98%, CAS: 97753-82-7, 100 mg
SDP
Catalog No. 502913545
Encompass_Preferred
A chromogenic substrate for β-galactosidase; upon enzymatic hydrolysis, it generates an indigo-colored precipitate that can be used to measure β-galactosidase activity; has been used to detect β-galactosidase in muscle from E. coli lacZ-expressing transgenic mice
Catalog No. 50-291-3545 Supplier Cayman Chemical Supplier No. 35299100 MG
A chromogenic substrate for β-galactosidase; upon enzymatic hydrolysis, it generates an indigo-colored precipitate that can be used to measure β-galactosidase activity; has been used to detect β-galactosidase in muscle from E. coli lacZ-expressing transgenic mice