Learn More
Cayman Chemical 5bromo4chLRo3Indolyl 10mg
SDP
Catalog No. 502946807
Encompass_Preferred
A chromogenic substrate for β-glucuronidase; upon cleavage by β-glucuronidase, an indigo pigment is released that can be used as a marker of β-glucuronidase activity
Catalog No. 50-294-6807 Supplier Cayman Chemical Supplier No. 3359110 MG
A chromogenic substrate for β-glucuronidase; upon cleavage by β-glucuronidase, an indigo pigment is released that can be used as a marker of β-glucuronidase activity