Learn More
Cayman Chemical 5bromo4chLRo3Indolyl 50mg
SDP
Catalog No. 502950690
Encompass_Preferred
A chromogenic substrate for β-glucuronidase; upon cleavage by β-glucuronidase, an indigo pigment is released that can be used as a marker of β-glucuronidase activity
Catalog No. 50-295-0690 Supplier Cayman Chemical Supplier No. 3359150 MG
A chromogenic substrate for β-glucuronidase; upon cleavage by β-glucuronidase, an indigo pigment is released that can be used as a marker of β-glucuronidase activity