Learn More
Cayman Chemical 5CarytetramthylrhodamIn 10mg
SDP
Catalog No. 502948258
Encompass_Preferred
A fluorescent dye; has been used for the covalent labeling of oligonucleotides for DNA analysis, fluorescence polarization assays, and protein FRET experiments; ex/em = 546/580 nm
Catalog No. 50-294-8258 Supplier Cayman Chemical Supplier No. 3459810 MG
A fluorescent dye; has been used for the covalent labeling of oligonucleotides for DNA analysis, fluorescence polarization assays, and protein FRET experiments; ex/em = 546/580 nm