Learn More
Cayman Chemical 5-Carboxytetramethylrhodamine succinimidyl ester, Purity: ≥90%, CAS: 150810-68-7, 10 mg
SDP
Catalog No. 502940075
Encompass_Preferred
An amine-reactive fluorescent probe; has been used in double site-directed peptide modification to label proteinase substrates for use in FRET assays; quenches lucifer yellow fluorescence by 90% in uncleaved proteinase substrates, which allows for the detection of enzyme-cleaved substrates by an increase in fluorescence intensity; has also been used in the synthesis of fluorescent derivatives of a variety of compounds, including ampicillin, UDP, and RU486; ex/em = 540-560/580 nm, respectively
Catalog No. 50-294-0075 Supplier Cayman Chemical Supplier No. 2850910 MG
An amine-reactive fluorescent probe; has been used in double site-directed peptide modification to label proteinase substrates for use in FRET assays; quenches lucifer yellow fluorescence by 90% in uncleaved proteinase substrates, which allows for the detection of enzyme-cleaved substrates by an increase in fluorescence intensity; has also been used in the synthesis of fluorescent derivatives of a variety of compounds, including ampicillin, UDP, and RU486; ex/em = 540-560/580 nm, respectively