Learn More
Cayman Chemical 5CLOro HydroCLOrothIazID 10mg
SDP
Catalog No. 502966812
Encompass_Preferred
A derivative of hydrochlorothiazide, which is a diuretic and antihypertensive agent that increases renal excretion of sodium, potassium, chloride, and bicarbonate ions by inhibiting tubular reabsorptive mechanisms
Catalog No. 50-296-6812 Supplier Cayman Chemical Supplier No. 1757110 MG
A derivative of hydrochlorothiazide, which is a diuretic and antihypertensive agent that increases renal excretion of sodium, potassium, chloride, and bicarbonate ions by inhibiting tubular reabsorptive mechanisms