Learn More
Cayman Chemical 5-Cyano-2,3-di-(p-tolyl)tetrazolium (chloride), CAS: 90217-02-0, 100 mg
SDP
Catalog No. 502970972
Encompass_Preferred
A redox-sensitive tetrazolium salt used primarily to detect metabolic activity in microorganisms; readily taken up by living cells and reduced by respiratory activity to produce a red fluorescent, water-insoluble formazan crystal (absorption maximum at 450 nm)
Catalog No. 50-297-0972 Supplier Cayman Chemical Supplier No. 15926100 MG
A redox-sensitive tetrazolium salt used primarily to detect metabolic activity in microorganisms; readily taken up by living cells and reduced by respiratory activity to produce a red fluorescent, water-insoluble formazan crystal (absorption maximum at 450 nm)