Learn More
Cayman Chemical 5'-Deoxy-5'-methylthioadenosine, Purity: ≥95%, CAS: 2457-80-9, 50 mg
SDP
Catalog No. 502968862
Encompass_Preferred
An intermediate in the generation of adenine and methionine that is produced by the decarboxylation of SAM; a potent agonist of adenosine receptors (Kis = 0.15, 1.13, 13.9, and 0.68 μM for A1, A2A, A2B, and A3, respectively); inhibits several enzymes at higher concentrations
Catalog No. 50-296-8862 Supplier Cayman Chemical Supplier No. 1559350 MG
An intermediate in the generation of adenine and methionine that is produced by the decarboxylation of SAM; a potent agonist of adenosine receptors (Kis = 0.15, 1.13, 13.9, and 0.68 μM for A1, A2A, A2B, and A3, respectively); inhibits several enzymes at higher concentrations