Learn More
Medchemexpress LLC (5E,9E,13E)-Teprenone | 3796-63-2 | 99.9% | 100 MG
SDP
Catalog No. 5000408793
Encompass_Preferred
(5E,9E,13E)-Teprenone is an isomer with antiulcer activity. It induces the transcriptional activation of HSP genes, which may enhance gastric mucosal defense under stress conditions.
  • Antiulcer activity.
  • Induces transcriptional activation of heat shock protein protein genes.
  • May increase gastric mucosal defense at conditions of stress.
  • Available in various quantities.
  • Purity of 99.9%.
Catalog No. 50-004-08793 Supplier Medchemexpress LLC Supplier No. HYB0779A100MG
(5E,9E,13E)-Teprenone is an isomer with antiulcer activity. It induces the transcriptional activation of HSP genes, which may enhance gastric mucosal defense under stress conditions.
  • Antiulcer activity.
  • Induces transcriptional activation of heat shock protein protein genes.
  • May increase gastric mucosal defense at conditions of stress.
  • Available in various quantities.
  • Purity of 99.9%.