Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (5E,9E,13E)-teprenone | 3796-63-2 | 99.9% | 330.56 g/mol | C23H38O | 10 MG
SDP

Catalog No. 5000296980
Encompass_Preferred

(5E,9E,13E)-teprenone is a geranylgeranyl-derived small molecule used in research to induce heat shock proteins (HSPs) and to study gastric mucosal protection. Supplied as a high-purity reagent suitable for biochemical and cellular assays.

  • High purity (99.9%) suitable for research applications.
  • CAS number 3796-63-2 for unambiguous identification.
  • Chemical formula C23H38O; molecular weight 330.56 g/mol.
  • Supplied as a 10 mg pack for small-scale experiments.
  • Recommended storage: 4°C; in solvent: -80°C (6 months) or -20°C (1 month) under nitrogen.

Catalog No. 50-002-96980 Supplier Medchemexpress LLC Supplier No. HYB0779A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(5E,9E,13E)-teprenone is a geranylgeranyl-derived small molecule used in research to induce heat shock proteins (HSPs) and to study gastric mucosal protection. Supplied as a high-purity reagent suitable for biochemical and cellular assays.

  • High purity (99.9%) suitable for research applications.
  • CAS number 3796-63-2 for unambiguous identification.
  • Chemical formula C23H38O; molecular weight 330.56 g/mol.
  • Supplied as a 10 mg pack for small-scale experiments.
  • Recommended storage: 4°C; in solvent: -80°C (6 months) or -20°C (1 month) under nitrogen.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.